missing translation for 'onlineSavingsMsg'
Learn More
Glycolic acid 70% in water, Thermo Scientific Chemicals Product Code.: 10475271

Glycolic acid 70% in water, Thermo Scientific Chemicals

Product Code. 10475271
1 kg, Glass bottle
Click to view available options
Quantity:
250 g
1 kg
25 kg
Packaging:
Glass bottle
Metal drum
This item is not returnable. View return policy

Product Code. 10475271

Brand: Thermo Scientific Acros 411030010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79-14-1, 7732-18-5
Molecular Formula C2H4O3
Molecular Weight (g/mol) 76.051
MDL Number MFCD00004312
InChI Key AEMRFAOFKBGASW-UHFFFAOYSA-N
Synonym glycolic acid, hydroxyacetic acid, glycollic acid, hydroxyethanoic acid, acetic acid, hydroxy, glycolate, caswell no. 470, alpha-hydroxyacetic acid, kyselina glykolova, kyselina glykolova czech
PubChem CID 757
ChEBI CHEBI:17497
IUPAC Name 2-hydroxyacetic acid
SMILES C(C(=O)O)O

Specifications

Melting Point 10.0°C
Boiling Point 113.0°C
Color Yellow
Linear Formula CH2OHCOOH
Formula Weight 76.04
Percent Purity 70%
Density 1.2700 g/mL
Quantity 1 kg
Packaging Glass bottle
Physical Form Solution
Specific Gravity 1.27
Chemical Name or Material Glycolic acid, 70% in water
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0462608 Certificate TypeCertificate of Analysis Date07/05/2024 Catalog Number411030010,411032500,411030250
Lot NumberA0461449 Certificate TypeCertificate of Analysis Date26/03/2024 Catalog Number411030010,411032500,411030250
Lot NumberA0460395 Certificate TypeCertificate of Analysis Date20/02/2024 Catalog Number411030010,411032500,411030250
Lot NumberA0457083 Certificate TypeCertificate of Analysis Date18/10/2023 Catalog Number411030010,411032500,411030250
Lot NumberA0454348 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number411030010,411032500,411030250
compliance-icons compliance-icons
Product Identifier
  • Glycolic acid, 67% in water
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Reproductive toxicity Category 1B
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H360FD-May damage fertility. May damage the unborn child.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Glycolic acid 70% in water, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.