Learn More
Glycolic acid, 98%, Thermo Scientific Chemicals
£38.79 - £125.63
Chemical Identifiers
CAS | 79-14-1 |
---|---|
Molecular Formula | C2H4O3 |
Molecular Weight (g/mol) | 76.051 |
MDL Number | MFCD00004312 |
InChI Key | AEMRFAOFKBGASW-UHFFFAOYSA-N |
Synonym | glycolic acid, hydroxyacetic acid, glycollic acid, hydroxyethanoic acid, acetic acid, hydroxy, glycolate, caswell no. 470, alpha-hydroxyacetic acid, kyselina glykolova, kyselina glykolova czech |
PubChem CID | 757 |
ChEBI | CHEBI:17497 |
IUPAC Name | 2-hydroxyacetic acid |
SMILES | C(C(=O)O)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11424164
|
Thermo Scientific Alfa Aesar
A12511.22 |
100 g |
£38.79
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11434164
|
Thermo Scientific Alfa Aesar
A12511.36 |
500 g |
£125.63
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Glycolic acid is used for acne and anti-aging treatment. It is also employed in the textile industry as a dyeing and tanning agent. It is used in emulsion polymers, solvents and additives for ink and paints. It is also useful as an intermediate for organic synthesis such as redox reactions, esterification and long chain polymerization. It acts as a monomer in the preparation of polyglycolic acid and other biocompatible copolymers. Further, it is used as a preservative and flavoring agent in organic synthesis. It is also used in adhesives and plastics.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsGlycolic acid is used for acne and anti-aging treatment. It is also employed in the textile industry as a dyeing and tanning agent. It is used in emulsion polymers, solvents and additives for ink and paints. It is also useful as an intermediate for organic synthesis such as redox reactions, esterification and long chain polymerization. It acts as a monomer in the preparation of polyglycolic acid and other biocompatible copolymers. Further, it is used as a preservative and flavoring agent in organic synthesis. It is also used in adhesives and plastics.
Solubility
Soluble in water, methanol, diethyl ether, acetone, acetic acid and ethyl acetate.
Notes
Hygroscopic. Incompatible with bases, oxidizing agents and reducing agents.
Chemical Identifiers
79-14-1 | |
76.051 | |
AEMRFAOFKBGASW-UHFFFAOYSA-N | |
757 | |
2-hydroxyacetic acid |
C2H4O3 | |
MFCD00004312 | |
glycolic acid, hydroxyacetic acid, glycollic acid, hydroxyethanoic acid, acetic acid, hydroxy, glycolate, caswell no. 470, alpha-hydroxyacetic acid, kyselina glykolova, kyselina glykolova czech | |
CHEBI:17497 | |
C(C(=O)O)O |
Specifications
2 | |
74°C to 80°C | |
C2H4O3 | |
MFCD00004312 | |
1209322 | |
glycolic acid, hydroxyacetic acid, glycollic acid, hydroxyethanoic acid, acetic acid, hydroxy, glycolate, caswell no. 470, alpha-hydroxyacetic acid, kyselina glykolova, kyselina glykolova czech | |
AEMRFAOFKBGASW-UHFFFAOYSA-N | |
2-hydroxyacetic acid | |
757 | |
76.05 | |
1.49 | |
Hygroscopic |
79-14-1 | |
98% | |
HOCH2CO2H | |
UN3261 | |
14,4498 | |
Soluble in water,methanol,diethyl ether,acetone,acetic acid and ethyl acetate. | |
C(C(=O)O)O | |
76.051 | |
CHEBI:17497 | |
98% | |
Unpleasant | |
Glycolic acid |
Safety and Handling
GHS H Statement
H314-H318-H302-H332
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful if inhaled.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H332-H335
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
missing translation for 'einecsNumber' : 201-180-5
missing translation for 'rtecsNumber' : MC5250000
missing translation for 'tsca' : Yes
missing translation for 'storageNote' : Hygroscopic
Recommended Storage : Ambient temperatures; Store under Nitrogen
- Glycolic acid
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H332-Harmful if inhaled.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : Hydroxyacetic acid
RUO – Research Use Only