missing translation for 'onlineSavingsMsg'
Learn More
Glycolic acid, 99%, Thermo Scientific Chemicals Product Code.: 15790809

Glycolic acid, 99%, Thermo Scientific Chemicals

Product Code. 15790809
25 g, Glass Bottle
Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg
Packaging:
Glass Bottle
This item is not returnable. View return policy

Product Code. 15790809

Brand: Thermo Scientific Acros 154510250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79-14-1
Molecular Formula C2H4O3
Molecular Weight (g/mol) 76.051
InChI Key AEMRFAOFKBGASW-UHFFFAOYSA-N
Synonym glycolic acid, hydroxyacetic acid, glycollic acid, hydroxyethanoic acid, acetic acid, hydroxy, glycolate, caswell no. 470, alpha-hydroxyacetic acid, kyselina glykolova, kyselina glykolova czechShow MoreShow Less
PubChem CID 757
ChEBI CHEBI:17497
IUPAC Name 2-hydroxyacetic acid
SMILES C(C(=O)O)O

Specifications

Melting Point 72.0°C to 80.0°C
Boiling Point 100.0°C
Color White
Assay Percent Range 99%
Linear Formula HOCH2CO2H
Beilstein 03, 228
Merck Index 15, 4534
Formula Weight 76.04
Percent Purity 99%
Quantity 25 g
Flash Point >300°C
Infrared Spectrum Conforms to Ref
Packaging Glass Bottle
Physical Form Adhering Crystals
Vapor Pressure 8.1mmHg at 80°C
Chemical Name or Material Glycolic acid
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0395325 Certificate TypeCertificate of Analysis Date25/11/2024 Catalog Number154515000,154510025,154510250,154511000
Lot NumberA0463910 Certificate TypeCertificate of Analysis Date26/06/2024 Catalog Number154515000,154510025,154510250,154511000
Lot NumberA0460343 Certificate TypeCertificate of Analysis Date20/02/2024 Catalog Number154515000,154510025,154510250,154511000
Lot NumberA0377542 Certificate TypeCertificate of Analysis Date17/10/2023 Catalog Number154515000,154510025,154510250,154511000
Lot NumberA0400593 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number154515000,154510025,154510250,154511000
compliance-icons compliance-icons
Product Identifier
  • Glycolic acid
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : 2-hydroxy-acetic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Glycolic acid, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.