missing translation for 'onlineSavingsMsg'
Learn More
Glycolic acid, ca 66-70% aq. soln., Thermo Scientific Chemicals Product Code.: 11348096

Glycolic acid, ca 66-70% aq. soln., Thermo Scientific Chemicals

Product Code. 11348096
500 mL
Click to view available options
Quantity:
500 mL
2500 mL
This item is not returnable. View return policy

Product Code. 11348096

Brand: Thermo Scientific Alfa Aesar L14980.AP

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Glycolic acid is used to evaluate the efficacy of glycolic peels treatment for all types of acne. It is also used in the fine synthesis of medicine and as a raw material of cosmetics and organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Glycolic acid is used to evaluate the efficacy of glycolic peels treatment for all types of acne. It is also used in the fine synthesis of medicine and as a raw material of cosmetics and organic synthesis.

Solubility
Soluble in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Store in cool place. Incompatible with bases and oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79-14-1
Molecular Formula C2H4O3
Molecular Weight (g/mol) 76.051
MDL Number MFCD00004312
InChI Key AEMRFAOFKBGASW-UHFFFAOYSA-N
Synonym glycolic acid, hydroxyacetic acid, glycollic acid, hydroxyethanoic acid, acetic acid, hydroxy, glycolate, caswell no. 470, alpha-hydroxyacetic acid, kyselina glykolova, kyselina glykolova czechShow MoreShow Less
PubChem CID 757
ChEBI CHEBI:17497
IUPAC Name 2-hydroxyacetic acid
SMILES C(C(=O)O)O

Specifications

Boiling Point 112°C
Linear Formula HOCH2CO2H
UN Number UN3265
Beilstein 1209322
Merck Index 14,4498
Solubility Information Soluble in water.
Concentration ≈67% aq. soln.
Formula Weight 76.05
Density 1.27
Quantity 500 mL
Flash Point None
Odor Sweet
Refractive Index 1.411
Chemical Name or Material Glycolic acid
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10242889 Certificate TypeCertificate of Analysis Date17/08/2022 Catalog NumberL14980.AP,L14980.0F
Lot Number10213730 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL14980.AP,L14980.0F
Lot Number10130479 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL14980.AP,L14980.0F
Lot Number10186898 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberL14980.AP,L14980.0F
Lot Number10222657 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberL14980.AP,L14980.0F

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Glycolic acid, ca 66-70% aq. soln., Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.