missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Glycylglycylglycine, 98+%
Click to view available options
Quantity:
1 g
5 g
25 g
Description
Glycylglycylglycine is used as a model peptide for studies of physicochemical parameters and molecular associations of small peptides. Triglycine is used as a copper chelator. It also acts as substrate for reproducible serum protein measurements by the biuret reaction1 and for the assay of aminotripeptidases
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Glycylglycylglycine is used as a model peptide for studies of physicochemical parameters and molecular associations of small peptides. Triglycine is used as a copper chelator. It also acts as substrate for reproducible serum protein measurements by the biuret reaction1 and for the assay of aminotripeptidases
Solubility
Soluble in water (94.6 mg/ml at 20°C), methanol (50 mg/ml), and 80% acetic acid (50 mg/ml).
Notes
Store at -20°C. Protect from moisture. Avoid excessive heat and light.
Glycylglycylglycine is used as a model peptide for studies of physicochemical parameters and molecular associations of small peptides. Triglycine is used as a copper chelator. It also acts as substrate for reproducible serum protein measurements by the biuret reaction1 and for the assay of aminotripeptidases
Solubility
Soluble in water (94.6 mg/ml at 20°C), methanol (50 mg/ml), and 80% acetic acid (50 mg/ml).
Notes
Store at -20°C. Protect from moisture. Avoid excessive heat and light.
Specifications
Melting Point | ∼240°C (decomposition) |
Beilstein | 1711130 |
Merck Index | 14,6579 |
Solubility Information | Soluble in water (94.6mg/ml at 20°C),methanol (50mg/ml),and 80% acetic acid (50mg/ml). |
Formula Weight | 191.14 |
Percent Purity | 99% |
Quantity | 1 g |
Chemical Name or Material | Glycylglycylglycine |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'