Learn More
Glyoxal, 40% w/w aq. soln., Thermo Scientific Chemicals
£19.40 - £183.00
Chemical Identifiers
CAS | 107-22-2 |
---|---|
Molecular Formula | C2H2O2 |
Molecular Weight (g/mol) | 58.036 |
MDL Number | MFCD00006957 |
InChI Key | LEQAOMBKQFMDFZ-UHFFFAOYSA-N |
Synonym | glyoxal, ethanedial, oxalaldehyde, 1,2-ethanedione, glyoxylaldehyde, biformal, biformyl, diformal, diformyl, aerotex glyoxal 40 |
PubChem CID | 7860 |
ChEBI | CHEBI:34779 |
IUPAC Name | oxaldehyde |
SMILES | C(=O)C=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11428900
|
Thermo Scientific Alfa Aesar
A16144.AE |
100 mL |
£19.40
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11453197
|
Thermo Scientific Alfa Aesar
A16144.AP |
500 mL |
£25.70
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11463197
|
Thermo Scientific Alfa Aesar
A16144.0F |
2500 mL |
£66.30
2500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11473197
|
Thermo Scientific Alfa Aesar
A16144.0D |
10,000 mL |
£183.00
10000mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Glyoxal is used to prepare 4,5-dihydroxy-2-imidazolidinone by condensation with urea. It finds application in leather tanning process, textile finishes and paper coatings. It is an important building block in the synthesis of imidazoles. It acts as a solubilizer and cross-linking agent in polymer chemistry. Further, it is used as a fixative for histology to preserve cells in order to examine under a microscope.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsGlyoxal is used to prepare 4,5-dihydroxy-2-imidazolidinone by condensation with urea. It finds application in leather tanning process, textile finishes and paper coatings. It is an important building block in the synthesis of imidazoles. It acts as a solubilizer and cross-linking agent in polymer chemistry. Further, it is used as a fixative for histology to preserve cells in order to examine under a microscope.
Solubility
Miscible with water, methanol and dimethylsulfoxide.
Notes
Moisture and heat sensitive. Incompatible with strong oxidizing agents, strong reducing agent, oxygen, peroxides, amides, amines, hydroxy-containing materials, nitric acid and aldehydes.
Chemical Identifiers
107-22-2 | |
58.036 | |
LEQAOMBKQFMDFZ-UHFFFAOYSA-N | |
7860 | |
oxaldehyde |
C2H2O2 | |
MFCD00006957 | |
glyoxal, ethanedial, oxalaldehyde, 1,2-ethanedione, glyoxylaldehyde, biformal, biformyl, diformal, diformyl, aerotex glyoxal 40 | |
CHEBI:34779 | |
C(=O)C=O |
Specifications
107-22-2 | |
1.271 | |
C2H2O2 | |
1.409 | |
1732463 | |
glyoxal, ethanedial, oxalaldehyde, 1,2-ethanedione, glyoxylaldehyde, biformal, biformyl, diformal, diformyl, aerotex glyoxal 40 | |
LEQAOMBKQFMDFZ-UHFFFAOYSA-N | |
oxaldehyde | |
7860 | |
58.04 | |
Glyoxal |
-14°C | |
104°C | |
HCOCHO | |
MFCD00006957 | |
14,4509 | |
Miscible with water,methanol and dimethylsulfoxide. | |
C(=O)C=O | |
58.036 | |
CHEBI:34779 | |
40% w/w aq. soln. |
Safety and Handling
GHS H Statement
H341-H315-H319-H317
Suspected of causing genetic defects.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
P201-P202-P261-P264b-P271-P272-P280-P281-P302+P352-P304+P340-P305+P351+P338-P308+P313-P333+P313-P362-P501c
H315-H317-H319-H332-H335-H341
missing translation for 'einecsNumber' : 203-474-9
missing translation for 'rtecsNumber' : MD2700000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Glyoxal, 40% w/w aq. soln
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Germ cell mutagenicity Category 2
- Skin corrosion/irritation Category 2
- Skin sensitiser Category 1
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H317-May cause an allergic skin reaction.
- H319-Causes serious eye irritation.
- H332-Harmful if inhaled.
- H335-May cause respiratory irritation.
- H341-Suspected of causing genetic defects.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : Ethanedial, Ethyleneglycol
RUO – Research Use Only