missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glyoxal-bis(2-hydroxyanil), ≥97.0% (from N), Honeywell Fluka™
Click to view available options
Quantity:
25 g
6 x 25 g
Packaging:
Glass Bottle
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
Melting Point | 203°C to 211°C |
UN Number | NONH for all modes of transport |
Beilstein | 2808815 |
IUPAC Name | 6-[(E)-2-(2-hydroxyanilino)ethenyl]iminocyclohexa-2,4-dien-1-one |
Formula Weight | 240.26g/mol |
Percent Purity | ≥97% (from N) |
Odor | Odourless |
Packaging | Glass Bottle |
Suitability | Passes test for suitable for metal definition |
Color | Yellow |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction