missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Glyoxal, pure, 40 wt.% solution in water

Brand:  Thermo Scientific Chemicals 156220250

Code : 29

Additional Details : CAS Number : 7732-18-5 Weight : 31.62500kg


 View more versions of this product

Product Code. 10593314

  • £388.00 / 25 L
Estimated Shipment: 31-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Glyoxal
107-22-2,7732-18-5
Colorless to Yellow
104.0°C
Metal Drum
HCOCHO
25 L
01,413
15, 4544
Solubility in water: miscible.
C(=O)C=O
58.04
8 mPa.s (20°C)
58.04
Pure
40 wt.% Solution in Water
-14.0°C
1.2650g/mL
>104°C
C2H2O2
MFCD00006957
01, 759
1.265
glyoxal, ethanedial, oxalaldehyde, 1,2-ethanedione, glyoxylaldehyde, biformal, biformyl, diformal, diformyl, aerotex glyoxal 40
LEQAOMBKQFMDFZ-UHFFFAOYSA-N
oxaldehyde
7860
CHEBI:34779
39 to 41% (Titrimetry other)
Liquid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Glyoxal, 40 wt% solution in water

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Germ cell mutagenicity Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2

Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H341-Suspected of causing genetic defects.
  • H332-Harmful if inhaled.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.

SDS Documents

Documents

Certificates
Special Offers

Special Offers

RUO – Research Use Only