Learn More
GMP Classic Lab Chair White
Brand: Techsit WS1220E/WHITE
Code : Z1
Additional Details : Weight : 12.15000kg
Description
Lab chairs for GMP areas that are suitable for S- classified laboratories. Closed GMP seat support cover made of plastic and chrome-plated polished metal parts, safety gas-spring protected by cover tube. All materials are washable and disinfectant resistant.• PUR integral foam backrest and seat.
• Chrome-plated cover tube.
• Height adjustment with safety gas spring.
• Polished diecast aluminum five-star base with glides.