Learn More
GMP Classic Lab High Chair white
Brand:  Techsit WS1211E/WHITE
Code : Z1
Additional Details : Weight : 11.95000kg
Product Code. 15484563
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
Lab chairs for GMP areas that are suitable for S- classified laboratories. Closed GMP seat support cover made of plastic and chrome-plated polished metal parts, safety gas-spring protected by cover tube. All materials are washable and disinfectant resistant.
• PUR integral foam backrest and seat.
• Chrome-plated cover tube.
• Height adjustment with safety gas spring.
• Polished diecast aluminum five-star base with glides.