missing translation for 'onlineSavingsMsg'
Learn More

Sampling Systems™ GMP Pallet Knives

Sampling Systems™ GMP Pallet Knives are a range of knives made from Stainless Steel that are ideal for hygienic applications.

Brand:  Sampling Systems™ A330-100

Additional Details : Weight : 0.01140kg

 View more versions of this product

Product Code. 15849621

  • £44.25 / Each
Estimated Shipment: 25-07-2024
to see stock.
Add to basket

Description

Description

  • Crevice free construction
  • All Stainless Steel construction
  • Satin finish
Specifications

Specifications

Pallet Knife
Polished
100 mm
Stainless Steel
185 mm
20 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers