missing translation for 'onlineSavingsMsg'
Learn More

Sampling Systems™ GMP Pallet Knives

Sampling Systems™ GMP Pallet Knives are a range of knives made from Stainless Steel that are ideal for hygienic applications.

Brand:  Sampling Systems™ A330-200

Additional Details : Weight : 0.02310kg

 View more versions of this product

Product Code. 15879621

  • £81.05 / Each
Estimated Shipment: 17-07-2024
to see stock.
Add to basket

Description

Description

  • Crevice free construction
  • All Stainless Steel construction
  • Satin finish
Specifications

Specifications

Pallet Knife
Polished
200 mm
Stainless Steel
325 mm
32 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers