missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GMPS Antibody (1D10), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00008833-M01
Additional Details : Weight : 0.00970kg
Description
GMPS Monoclonal antibody specifically detects GMPS in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), ELISASpecifications
GMPS | |
Monoclonal | |
Unconjugated | |
NP_003866 | |
Mouse | |
IgG purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Sandwich ELISA | |
1D10 | |
In 1x PBS, pH 7.4 | |
EC 6.3.5.2, Glutamine amidotransferase, GMP synthase, GMP synthase [glutamine-hydrolyzing], GMP synthetase, guanine monphosphate synthetase, guanosine 5'-monophosphate synthase, MLL/GMPS fusion protein | |
GMPS (NP_003866, 108 a.a. ~ 215 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. QMMNKVFGGTVHKKSVREDGVFNISVDNTCSLFRGLQKEEVVLLTHGDSVDKVADGFKVVARSGNIVAGIANESKKLYGAQFHPEVGLTENGKVILKNFLYDIAGCSG | |
0.1 mg | |
metabolism | |
8833 | |
Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
IgG1 κ |