missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GMPS Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Bio-Techne NBP2-56670
Additional Details : Weight : 0.00970kg
Description
GMPS Polyclonal specifically detects GMPS in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
GMPS | |
Polyclonal | |
Immunocytochemistry/Immunofluorescence 1-4 ug/ml | |
EC 6.3.5.2, Glutamine amidotransferase, GMP synthase, GMP synthase [glutamine-hydrolyzing], GMP synthetase, guanine monphosphate synthetase, guanosine 5'-monophosphate synthase, MLL/GMPS fusion protein | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
GMPS | |
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:AGGDLKDGHHHYEGAVVILDAGAQYGKVIDRRVRELFVQSEIFPLETPAFAIKEQGFRAIIISGGPNSVYAEDAPWFDPAIFTIGKPVLGICYGMQMMNKVFGGTVHKKSVREDGVFNISVDNTCSLFRGLQKEEVVLLT | |
100 μL | |
metabolism | |
8833 | |
Human | |
IgG |
For Research Use Only