missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GMPS Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Bio-Techne NBP2-16696
Additional Details : Weight : 0.00970kg
Description
GMPS Polyclonal specifically detects GMPS in Human samples. It is validated for Western Blot.Specifications
GMPS | |
Polyclonal | |
Unconjugated | |
0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal | |
EC 6.3.5.2, Glutamine amidotransferase, GMP synthase, GMP synthase [glutamine-hydrolyzing], GMP synthetase, guanine monphosphate synthetase, guanosine 5'-monophosphate synthase, MLL/GMPS fusion protein | |
Rabbit | |
77 kDa | |
0.1 mL | |
metabolism | |
8833 | |
Human | |
IgG |
Western Blot | |
0.69mg/mL | |
Western Blot 1:500-1:3000 | |
P49915 | |
GMPS | |
Recombinant protein encompassing a sequence within the center region of human GMP synthase. The exact sequence is proprietary. | |
Affinity Purified | |
RUO | |
Primary | |
Human. Expected to cross react based on sequence identity: Mouse (96%), Chicken (91%), Rat (97%), Zebrafish (85%), Bovine (97%). | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
For Research Use Only