missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GMPS, Mouse anti-Human, Clone: 1D10, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00008833-M01.100ug
Additional Details : Weight : 0.00010kg
Description
Sequence: QMMNKVFGGTVHKKSVREDGVFNISVDNTCSLFRGLQKEEVVLLTHGDSVDKVADGFKVVARSGNIVAGIANESKKLYGAQFHPEVGLTENGKVILKNFLYDIAGCSGSpecifications
GMPS | |
Monoclonal | |
Unconjugated | |
guanine monphosphate synthetase | |
- | |
Mouse | |
100 μg | |
Primary | |
Human | |
Antibody |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
1D10 | |
In 1x PBS, pH 7.4 | |
NM_003875 | |
GMPS | |
GMPS (NP_003866, 108 a.a. ∼ 215 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
RUO | |
8833 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG1 κ |