missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GMPS, Mouse anti-Human, Polyclonal Antibody, Abnova™
Mouse Polyclonal Antibody
Brand: Abnova H00008833-B01P.50ug
Additional Details : Weight : 0.00010kg
Description
Sequence: MALCNGDSKLENAGGDLKDGHHHYEGAVVILDAGAQYGKVIDRRVRELFVQSEIFPLETPAFAIKEQGFRAIIISGGPNSVYAEDAPWFDPAIFTIGKPVLGICYGMQMMNKVFGGTVHKKSVREDGVFNISVDNTCSLFRGLQKEEVVLLTHGDSVDKVADGFKVVARSGNIVAGIANESKKLYGAQFHPEVGLTENGKVILKNFLYDIAGCSGTFTVQNRELECIREIKERVGTSKVLVLLSGGVDSTVCTALLNRALNQEQVIAVHIDNGFMRKRESQSVEEALKKLGIQVKVINAAHSFYNGTTTLPISDEDRTPRKRISKTLNMTTSPEEKRKIIGDTFVKIANEVIGEMNLKPEEVFLAQGTLRPDLIESASLVASGKAELIKTHHNDTELIRKLREEGKVIEPLKDFHKDEVRILGRELGLPEELVSRHPFPGPGLAIRVICAEEPYICKDFPETNNILKIVADFSASVKKPHTLLQRVKACTTEEDQEKLMQITSLHSLNAFLLPIKTVGVQGDCRSYSYVCGISSKDEPDWESLIFLARLIPRMCHNVNRVVYIFGPPVKEPPTDVTPTFLTTGVLSTLRQADFEAHNILRESGYAGKISQMPVILTPLHFDRDPLQKQPSCQRSVVIRTFITSDFMTGIPATPGNEIPVEVVLKMVTEIKKIPGISRIMYDLTSKPPGTTEWESpecifications
GMPS | |
Polyclonal | |
In 1x PBS, pH 7.4 | |
NM_003875.2 | |
GMPS | |
GMPS (NP_003866.1, 1 a.a. ∼ 693 a.a) full-length human protein. | |
RUO | |
8833 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Purified |
Western Blot | |
Unconjugated | |
guanine monphosphate synthetase | |
- | |
Mouse | |
50 μg | |
Primary | |
Human | |
Antibody |