Learn More
Novus Biologicals™ GMPS Overexpression Lysate
Brand: Novus Biologicals™ NBL1-11151
Additional Details : Weight : 0.00970kg
Description
The GMPS Overexpression Lysate is derived from human. It has been validated for the following applications: Western Blot.Specifications
Western Blot | |
0.1 mg | |
Protein | |
-80°C. Avoid freeze-thaw cycles. | |
Native |
The lysate was created in HEK293T cells, using plasmid ID RC24267 and based on accession number NM_3875. The protein contains a C-terminal DDK tag. | |
RIPA buffer | |
Homo sapiens guanine monphosphate synthetase (GMPS), mRNA. | |
metabolism |
For Research Use Only