missing translation for 'onlineSavingsMsg'
Learn More

GMPS Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 16376-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16805984

  • £130.00 / 20µL
Estimated Shipment: 01-08-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Transcription factor which plays a key role in defining the composition of the RNA polymerase II (RNAPII) elongation complex and in modulating the production of mature mRNA transcripts. Acts as an assembly factor to recruit various factors to the RNAPII elongation complex and is recruited to the complex via binding to the transcription elongation factor SUPT6H bound to the C-terminal domain (CTD) of the RNAPII subunit RPB1 (POLR2A). The SUPT6H:IWS1:CTD complex recruits mRNA export factors (ALYREF/THOC4, EXOSC10) as well as histone modifying enzymes (such as SETD2) to ensure proper mRNA splicing, efficient mRNA export and elongation-coupled H3K36 methylation, a signature chromatin mark of active transcription.
Specifications

Specifications

GMPS
Polyclonal
Unconjugated
GMPS
Glutamine amidotransferase, GMP synthetase, GMPS
Rabbit
Antigen Affinity Chromatography
RUO
229363, 295088, 8833
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation
0.34 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P49915, Q3THK7, Q4V7C6
Gmps
GMPS Fusion Protein Ag9350
20 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers