missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva (formerly Pall Lab) GN-6 Membrane, 0.45 μm
Certified 0.45 μm membrane for microbiological analysis meets or surpasses regulatory specifications.
Brand: Cytiva (formerly Pall Lab) 66068
This item is not returnable.
View return policy
Description
- Designed for laboratory applications only
- Not approved for use in medical, clinical, surgical or other patient applications
Specifications
White | |
Metricel Mixed Cellulose Ester | |
5 x 200/Pk. | |
Gridded |
Bulk with Parchment Separators | |
0.45 μm | |
Gamma Irradiated | |
47 mm |