missing translation for 'onlineSavingsMsg'
Learn More

Cytiva (formerly Pall Lab) GN-6 Membrane, 0.45 μm

Certified 0.45 μm membrane for microbiological analysis meets or surpasses regulatory specifications.

Brand:  Cytiva (formerly Pall Lab) 66191

 View more versions of this product

Product Code. 17154551

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Designed for laboratory applications only
  • Not approved for use in medical, clinical, surgical or other patient applications
TRUSTED_SUSTAINABILITY
Specifications

Specifications

White
Metricel Mixed Cellulose Ester
10 x 200/Pk.
Gridded
Bulk with Parchment Separators
0.45 μm
Gamma Irradiated
47 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers