missing translation for 'onlineSavingsMsg'
Learn More

Cytiva (formerly Pall Lab) GN-6 Membrane, 0.45 μm

Certified 0.45 μm membrane for microbiological analysis meets or surpasses regulatory specifications.

Brand:  Cytiva (formerly Pall Lab) 66278

 View more versions of this product

Product Code. 17184551

  • £226.00 / Pack of 200
Estimated Shipment: 24-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Designed for laboratory applications only
  • Not approved for use in medical, clinical, surgical or other patient applications
TRUSTED_SUSTAINABILITY
Specifications

Specifications

White
Metricel Mixed Cellulose Ester
200/Pk.
Gridded
Bulk with Parchment Separators
0.45 μm
Gamma Irradiated
47 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers