Learn More
Novacyt - Microgen Bioproducts GN-ID A 12 Substrate ID System
Microgen Bioproducts GN-ID A - For the identification of aerobic and facultative anaerobic gram negative bacteria.
Brand:  Novacyt - Microgen Bioproducts MID-64
Product Code. 17383211
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Microgen GN-ID A is a convenient 12 substrate ID system for the most commonly isolated Enterobacteriaceae.
  • The Microgen GN-ID A System is for the identification of the commonly encountered Enterobacteriaceae from food or medical samples including urinary samples and common wound isolates.
  • For wider clinical applications and for veterinary and industrial samples, identification of all currently recognised species of Enterobacteriaceae and an extensive range of oxidase-positive Gram Negative Bacilli is achieved with the addition of the GN-ID Bidentification system.
Specifications
Specifications
ISO13485 ISO9001
Yes
Gelatin, Malonate, Inositol, Sorbitol, Rhamnose, Sucrose, Lactose, Arabinose, Adonitol, Raffinose, Salicin, Arginine, TDA, Nitrate
Bacterial Isolate
Test Strip
60 A Test strips
Monera
60 A Test strips
>99%
18 to 24 hours incubation for Enterobacteriaceae, and after 48 hours for oxidase positive isolates
2°C to 8°C
Non-waived
60 A Test strips
>99%
An Identification System for All Currently Recognised Enterobacteriaceae and an Extensive Range of Oxidase-Positive Gramnegative Bacilli
12/Test Strip
Carton with pouches and strip holding tray
All currently recognised Enterobacteriaceae and an extensive range of oxidase-positive Gramnegative Bacilli
Bacteria