Learn More
GNL3 Mouse anti-Human, Unconjugated, Clone: 3B8F10, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67169-1-IG-150UL
This item is not returnable.
View return policy
Description
The GNL3 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human GNL3. This antibody recognizes human, mouse, rat antigen. The GNL3 antibody has been validated for the following applications: IF, WB, ELISA analysis.
Specifications
GNL3 | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human, Rat, Mouse |
Western Blot, Immunofluorescence, ELISA | |
3B8F10 | |
Unconjugated | |
150 μL | |
26354 | |
IgG2a |