Learn More
GNMT Mouse anti-Human, Unconjugated, Clone: 1B5E3, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67294-1-IG-150UL
This item is not returnable.
View return policy
Description
The GNMT antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human GNMT. This antibody recognizes human, mouse, pig, rat antigen. The GNMT antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
GNMT | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human, Mouse, Rat, Pig |
Western Blot, Immunohistochemistry, ELISA | |
1B5E3 | |
Unconjugated | |
150 μL | |
27232 | |
IgG1 |