missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Armenian Hamster IgG (H+L) Secondary Antibody, HRP, Invitrogen™
Goat Polyclonal Secondary Antibody
Brand: Invitrogen PA132045
Code : LE
Additional Details : Weight : 0.01000kg
Description
Greatly diminished reactivity will occur against Golden Syrian hamster IgG.
Anti-Hamster secondary antibodies are affinity-purified antibodies with well-characterized specificity for hamster immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.Specifications
Armenian Hamster IgG (H+L) | |
Polyclonal | |
HRP | |
Goat | |
Antigen affinity chromatography | |
RUO | |
Hamster | |
Liquid |
ELISA, Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 10mg/mL BSA and 0.01% gentamicin sulfate; pH 7.2 | |
Armenian Hamster IgG whole molecule | |
500 μg | |
Secondary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
IgG |