missing translation for 'onlineSavingsMsg'
Learn More

Goat anti-Human IgG Fc Recombinant Secondary Antibody, Alexa Fluor™ 488, Invitrogen™

Goat Recombinant Monoclonal Secondary Antibody

£88.75 - £277.00

Specifications

Antigen Human IgG Fc
Clone 3H8L9
Concentration 1.0 mg/mL
Applications Flow Cytometry, Immunocytochemistry, Western Blot
Classification Recombinant Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17876752
View Documents
Invitrogen
A5574750UL
50 μL
£88.75
50µL
Please to purchase this item. Need a web account? Register with us today!
17895972
View Documents
Invitrogen
A55747
250 μL
£277.00
250µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Goat anti-Human IgG Fc Recombinant Secondary Antibody specifically detects human IgG Fc with no cross-reactivity to human IgA or IgM or various species immunoglobulins. The sensitivity and specificity of each lot is confirmed using ELISA. Recombinant antibodies are produced using specific genes that code for the desired antibodies. These genes are cloned into an expression vector and expressed in vitro. The advantages of recombinant antibodies include: better specificity, animal origin-free formulation, and lot-to-lot consistency. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

Anti-Human secondary antibodies are affinity-purified antibodies with well-characterized specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Specifications

Specifications

Human IgG Fc
1.0 mg/mL
Recombinant Monoclonal
Liquid
RUO
PBS with 5mM sodium azide; pH 7.4
IgG
Protein G
Secondary Antibody
3H8L9
Flow Cytometry, Immunocytochemistry, Western Blot
Alexa Fluor 488
Goat
Human
Purified Human IgG
Secondary
4° C, store in dark, DO NOT FREEZE!
Videos
SDS
Documents

Documents

Product Certifications