Learn More
Goat anti-Human IgG1 (Heavy chain) Recombinant Secondary Antibody, HRP, Invitrogen™
Goat Recombinant Monoclonal Secondary Antibody
£92.00 - £287.00
Specifications
Antigen | Human IgG1 (Heavy chain) |
---|---|
Clone | 3H60L9 |
Concentration | 1.0 mg/mL |
Applications | ELISA, Peptide Array, Western Blot |
Classification | Recombinant Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17888801
|
Invitrogen
A5573950UL |
50 μL |
£92.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17848801
|
Invitrogen
A55739 |
250 μL |
£287.00
250µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Goat anti-Human IgG1 (Heavy chain) Recombinant Monoclonal Antibody specifically detects human IgG1 heavy chain with no cross-reactivity to human IgG2, IgG3, IgG4, IgA or IgM or various species immunoglobulins, including monkey IgG. The sensitivity and specificity of each lot is confirmed using ELISA. Recombinant antibodies are produced using specific genes that code for the desired antibodies. These genes are cloned into an expression vector and expressed in vitro. The advantages of recombinant antibodies include: better specificity, animal origin-free formulation, and lot-to-lot consistency. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.
Anti-Human secondary antibodies are affinity-purified antibodies with well-characterized specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.Specifications
Human IgG1 (Heavy chain) | |
1.0 mg/mL | |
Recombinant Monoclonal | |
Liquid | |
RUO | |
PBS with proprietary stabilizer and proprietary preservative; pH 7.4 | |
IgG | |
Protein G | |
Secondary Antibody |
3H60L9 | |
ELISA, Peptide Array, Western Blot | |
HRP | |
Goat | |
Human | |
Purified Human IgG | |
Secondary | |
4° C, do not freeze |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.