missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Mouse Ig, Polyclonal, Secondary Antibody, BD
Brand: BD Biosciences 553998
Additional Details : Weight : 0.50000kg
Description
X
Host Species: Goat
Clone: Polyclonal
Isotype: Ig
Species Reactivity: Mouse
Flow Cytometry
Specifications
Goat anti-Mouse Ig | |
POLYCLONAL | |
Goat | |
RUO | |
Secondary | |
Ig |
Polyclonal | |
0.5mg/mL | |
0.5 mg | |
Immunology | |
Mouse |