Learn More
Invitrogen™ Goat anti-Mouse IgG (H+L) Secondary Antibody, DyLight™ 594
Goat Polyclonal Secondary Antibody
Brand: Invitrogen™ 35510
Description
Product # 35510 has been successfully used in Western blot, IF, ICC, IHC, IP and FACS applications. Product # 35510 reacts with the heavy chains of mouse IgG and with the light chains common to most mouse immunoglobulins, but does not react against non-immunoglobulin serum proteins. However, this antibody may cross-react with immunoglobulins from other species. Store product protected from light at 4°C until opened. To extend the shelf-life of this product, add an equal volume of glycerol to make a final concentration of approximately 50% glycerol and store at -20°C. DyLight 594 Amax= 593 nm; Emax= 618 nm. Mole Dye/Mole Protein Ratio is lot-dependent.
Anti-Mouse secondary antibodies are affinity-purified antibodies with well-characterized specificity for mouse immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Specifications
| Mouse IgG (H+L) | |
| Polyclonal | |
| DyLight 594 | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| Mouse | |
| Secondary Antibody | |
| IgG |
| Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 1% BSA and 0.02% sodium azide; pH 7.2 | |
| Purified Mouse IgG, whole molecule. | |
| 1 mL | |
| Secondary | |
| 4°C | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.