missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Mouse IgM Secondary Antibody, PE, Invitrogen™
Goat Polyclonal Secondary Antibody
Brand: Invitrogen PA186003
Code : LD
Additional Details : Weight : 0.01000kg
Description
Anti-Mouse secondary antibodies are affinity-purified antibodies with well-characterized specificity for mouse immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.Specifications
Mouse IgM | |
Polyclonal | |
PE | |
Goat | |
Affinity chromatography | |
RUO | |
Mouse | |
Liquid |
Flow Cytometry | |
0.5 mg/mL | |
PBS with sucrose and <0.1% sodium azide | |
Mouse IgM paraproteins. | |
500 μg | |
Secondary | |
4° C, do not freeze | |
IgG |