missing translation for 'onlineSavingsMsg'
Learn More

Goat anti-Rabbit IgG (H+L) Cross-Adsorbed ReadyProbes™ Secondary Antibody, Alexa Fluor™ 488, Invitrogen™

Goat Polyclonal Secondary Antibody

Brand:  Invitrogen R37116

Code : NEW

Additional Details : Weight : 0.02000kg

Product Code. 15196886

  • £176.00 / 2 ampoules
Estimated Shipment: 30-07-2024
to see stock.

Description

Description

No pipetting or dilutions required. Just tip and drip two drops per mL buffer to stain your cells.

Rabbit IgG (H+L) Cross-Adsorbed Polyclonal antibody specifically detects Rabbit IgG (H+L) Cross-Adsorbed in Rabbit samples. It is validated for Flow Cytometry, Immunocytochemistry.
Specifications

Specifications

Rabbit IgG (H+L) Cross-Adsorbed
Polyclonal
PBS with 0.1% BSA and <0.1% sodium azide
Gamma Immunoglobin
2 x 2.5 mL
Secondary
4° C, store in dark
IgG
Flow Cytometry, Immunocytochemistry
Alexa Fluor 488
Goat
Affinity chromatography
RUO
Rabbit
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers