missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Rat Ig, Polyclonal, biotin, Secondary Antibody, BD
Brand: BD Biosciences 554014
Additional Details : Weight : 0.01000kg
Description
Host Species: Goat
Clone: Polyclonal
Isotype: Ig
Species Reactivity: Rat
ELISA, Flow Cytometry
Specifications
Goat anti-Rat Ig | |
POLYCLONAL | |
Biotin | |
0.5 mg | |
Immunology | |
Rat |
Polyclonal | |
0.5mg/mL | |
Goat | |
RUO | |
Secondary | |
Ig |