Learn More
Goat Anti-Rat Ig, Polyclonal , PE, Secondary Antibody, BD
Brand: BD Biosciences 550767
Additional Details : Weight : 0.01000kg
Description
The polyclonal antibody was purified from antiserum by negative adsorption and affinity chromatography. The antibody was conjugated to APC under optimum conditions, and unconjugated antibody and free APC were removed. Store undiluted at 4°C and protec
Host Species: Goat
Isotype: Ig
Species Reactivity: Rat
Flow Cytometry
Specifications
Goat Anti-Rat Ig | |
POLYCLONAL | |
PE | |
0.2 mg | |
Immunology | |
Rat |
Monoclonal | |
0.2mg/mL | |
Goat | |
RUO | |
Secondary | |
Ig |