missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Rat IgG2a Secondary Antibody, HRP, Invitrogen™
Goat Polyclonal Secondary Antibody
Brand: Invitrogen PA184709
This item is not returnable.
View return policy
Description
Goat anti Rat IgG2a antibody recognizes rat IgG2a as assessed by immunoelectrophoresis and ELISA.Goat anti Rat IgG2a antibody may cross react with IgG2a from other species.
Anti-Rat secondary antibodies are affinity-purified antibodies with well-characterized specificity for rat immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.Specifications
Rat IgG2a | |
Polyclonal | |
HRP | |
Goat | |
Affinity chromatography | |
RUO | |
Rat | |
Liquid |
ELISA, Western Blot | |
1 mg/mL | |
PBS with 0.2% BSA and 0.05% ProClin 300 | |
Proprietary Immunogen. | |
500 μg | |
Secondary | |
4° C, do not freeze | |
IgG2a |