Learn More
Goat anti-Syrian/Armenian Hamster IgG (H+L) Cross-Adsorbed Secondary Antibody, FITC, Invitrogen™
Goat Polyclonal Secondary Antibody
Brand: Invitrogen PA533285
Code : NEW
Additional Details : Weight : 0.01000kg
Description
PA5-33285 targets Hamster IgG in FACS applications. The antibody has been cross adsorbed against both mouse and rat immunoglobulins to minimize cross-reactivity For FACS analysis, use 50 μL of the suggested working dilution to label 1x10^6 cells in 100 μL.
Anti-Hamster secondary antibodies are affinity-purified antibodies with well-characterized specificity for hamster immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.Specifications
Syrian/Armenian Hamster IgG (H+L) Cross-Adsorbed | |
Polyclonal | |
FITC | |
Goat | |
purified | |
RUO | |
Hamster | |
Liquid |
Flow Cytometry | |
0.7 mg/mL | |
PBS with 1% BSA and 0.09% sodium azide | |
Hamster IgG | |
700 μg | |
Secondary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark. | |
IgG |