missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Syrian Hamster IgG (H+L) Cross-Adsorbed Secondary Antibody, Alexa Fluor™ 488, Invitrogen™
Goat Polyclonal Secondary Antibody
Brand: Invitrogen A21110
Code : 50
Additional Details : Weight : 0.10000kg
Description
Product will be shipped at Room Temperature.
Syrian Hamster IgG (H+L) Cross-Adsorbed Polyclonal antibody specifically detects Syrian Hamster IgG (H+L) Cross-Adsorbed in Hamster samples. It is validated for Immunocytochemistry, Immunohistochemistry, Western Blot.Specifications
Syrian Hamster IgG (H+L) Cross-Adsorbed | |
Polyclonal | |
Alexa Fluor 488 | |
Goat | |
purified | |
RUO | |
Hamster | |
Liquid |
Immunocytochemistry, Immunohistochemistry, Western Blot | |
2 mg/mL | |
PBS with 5mM sodium azide; pH 7.5 | |
Gamma Immunoglobins Heavy and Light chains | |
1 mg | |
Secondary | |
4° C, store in dark | |
IgG |