missing translation for 'onlineSavingsMsg'
Learn More
Gold Chloride Trihydrate ACS, MP Biomedicals™ Product Code.: 15702262
compliance-icons compliance-icons

Gold Chloride Trihydrate ACS, MP Biomedicals™

Product Code. 15702262 Shop All MP Biomedicals Products
1 g
Click to view available options
Quantity:
1 g
This item is not returnable. View return policy

Product Code. 15702262

Brand: MP Biomedicals 0215252601

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MP Biomedicals™ Gold Chloride Trihydrate ACS for microanalysis of rubidium, cesium and in the determination of alkaloids.

  • Used for microanalysis of rubidium, cesium and in the determination of alkaloids.
  • Usually used as a harbinger for the preparation of gold (Au) nanoparticles.
  • It is used in the control rods of nuclear reactors and in the manufacturing of electric light filaments.
TRUSTED_SUSTAINABILITY

Specifications

CAS 16961-25-4
Quantity 1 g
Molecular Formula AuCl4H
Synonym Tetrachlorogold (III) Hydrate
InChI Key VDLSFRRYNGEBEJ-UHFFFAOYSA-K
SMILES Cl.Cl[Au](Cl)Cl
IUPAC Name trichlorogold hydrochloride
Molecular Weight (g/mol) 339.77
Formula Weight 393.8
Grade ACS
Chemical Name or Material Gold Chloride, Trihydrate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Gold Chloride Trihydrate, ACS
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Gold Chloride Trihydrate ACS, MP Biomedicals™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.