missing translation for 'onlineSavingsMsg'
Learn More
Gold foil, 0.1mm (0.004in) thick, Premion™, 99.9975+% (metals basis), Thermo Scientific Chemicals Product Code.: 11341189

Gold foil, 0.1mm (0.004in) thick, Premion™, 99.9975+% (metals basis), Thermo Scientific Chemicals

Product Code. 11341189
25 x 25 mm
Click to view available options
Quantity:
25 x 25 mm
50 x 50 mm
100 x 100 mm
This item is not returnable. View return policy

Product Code. 11341189

Brand: Thermo Scientific Alfa Aesar 011391.FF

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Form: approx. 1.2g/25x25mm

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-57-5
Molecular Formula Au
Molecular Weight (g/mol) 196.97
MDL Number MFCD00003436
InChI Key PCHJSUWPFVWCPO-UHFFFAOYSA-N
Synonym colloidal, powder, flake, leaf, gold, colloidal, burnish, shell, ci pigment metal 3, magnesium purple, c.i. pigment metal 3
PubChem CID 23985
ChEBI CHEBI:30050
IUPAC Name gold
SMILES [Au]

Specifications

Color Golden
Appearance Gold colored foil
Density 19.32g/cm3 at 20°C
Assay Percent Range >99.9975% (Metals basis)
Quantity 25 x 25 mm
Odor Odorless
Form Foil
Chemical Name or Material Gold

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberU15J021 Certificate TypeCertificate of Analysis Date24/10/2023 Catalog Number011391.FF,011391.GH,011391.FI
Lot NumberY02I023 Certificate TypeCertificate of Analysis Date15/11/2022 Catalog Number011391.FF,011391.GH,011391.FI
Lot NumberU11I041 Certificate TypeCertificate of Analysis Date19/08/2022 Catalog Number011391.FF,011391.GH,011391.FI
Lot NumberL21X023 Certificate TypeCertificate of Analysis Date17/11/2021 Catalog Number011391.FF,011391.GH,011391.FI
Lot NumberA23L17 Certificate TypeCertificate of Analysis Date17/11/2021 Catalog Number011391.FF,011391.GH,011391.FI

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Gold foil, 0.1mm (0.004in) thick, Premion™, 99.9975+% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.