missing translation for 'onlineSavingsMsg'
Learn More
Gold(I) chloride, 99+%, Thermo Scientific Chemicals Product Code.: 10571124

Gold(I) chloride, 99+%, Thermo Scientific Chemicals

Product Code. 10571124
5 g, Glass bottle
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10571124

Brand: Thermo Scientific Acros 388330050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10294-29-8
Molecular Formula AuCl
Molecular Weight (g/mol) 232.42
MDL Number MFCD00046175
InChI Key FDWREHZXQUYJFJ-UHFFFAOYSA-M
Synonym gold i chloride, gold monochloride, gold chloride, aurous chloride, gold chloride trihydride, acmc-1bvpa, gold monochloride, acid, gold i chloride trace metals basis, gold i chloride, saj special gradeShow MoreShow Less
PubChem CID 27366
ChEBI CHEBI:30078
IUPAC Name chlorogold
SMILES [Cl-].[Au+]

Specifications

Melting Point 289.0°C
Color Yellow to Green
Physical Form Powder
Quantity 5 g
Assay Percent Range 99+%
Linear Formula AuCl
Merck Index 15, 4550
Solubility Information Solubility in water: decomposes
Formula Weight 232.42
Percent Purity ≥99%
Packaging Glass bottle
Chemical Name or Material Gold(I) chloride
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0464259 Certificate TypeCertificate of Analysis Date16/07/2024 Catalog Number388330050,388330010
Lot NumberA0448633 Certificate TypeCertificate of Analysis Date25/12/2023 Catalog Number388330050,388330010
Lot NumberA0453757 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number388330050,388330010
Lot NumberA0405976 Certificate TypeCertificate of Analysis Date12/12/2022 Catalog Number388330050,388330010
Lot NumberA0443717 Certificate TypeCertificate of Analysis Date14/07/2022 Catalog Number388330050,388330010

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Gold(I) chloride, 99+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.