missing translation for 'onlineSavingsMsg'
Learn More

Gold(I) chloride, Premion™, 99.99% (metals basis), Au 84.2% min

Product Code. 11304038
Click to view available options
Quantity:
0.25 g
1 g
5 g
Unit Size:
0.25g
1g
5g
This item is not returnable. View return policy

Product Code. 11304038

Brand: Thermo Scientific Alfa Aesar 040432.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10294-29-8 | AuCl | 232.42 g/mol

As intermediates. Also In Chemicals, pharmaceutical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10294-29-8
Molecular Formula AuCl
Molecular Weight (g/mol) 232.42
MDL Number MFCD00046175
InChI Key FDWREHZXQUYJFJ-UHFFFAOYSA-M
Synonym gold i chloride, gold monochloride, gold chloride, aurous chloride, gold chloride trihydride, acmc-1bvpa, gold monochloride, acid, gold i chloride trace metals basis, gold i chloride, saj special grade
PubChem CID 27366
ChEBI CHEBI:30078
IUPAC Name chlorogold
SMILES [Cl-].[Au+]

Specifications

Melting Point 289°C (decomposition)
Physical Form Powder
Quantity 1 g
UN Number UN3260
Merck Index 14,4515
Solubility Information Very slightly soluble in water. soluble in HCl,HBr organic solvents.
Formula Weight 232.42
Percent Purity 99.99%
Grade Premion
Concentration or Composition (by Analyte or Components) Au 84.2% min.
Odor Odorless
Sensitivity Moisture sensitive
Density 7.57 g/mL
Assay (metals basis)
Chemical Name or Material Gold(I) chloride
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.