missing translation for 'onlineSavingsMsg'
Learn More

Gold(I) iodide, 99%, Thermo Scientific Chemicals

Catalog Number 11307198
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
10294-31-2
AuI
323.87
MFCD00014175
ATGIETUGWDAYPU-UHFFFAOYSA-M
gold monoiodide, gold i iodide, gold iodide, acmc-1bqvl, gold i iodide trace metals basis
82526
iodogold
[I-].[Au+]
This item is not returnable. View return policy
10294-31-2
AuI
323.87
MFCD00014175
ATGIETUGWDAYPU-UHFFFAOYSA-M
gold monoiodide, gold i iodide, gold iodide, acmc-1bqvl, gold i iodide trace metals basis
82526
iodogold
[I-].[Au+]

Gold(I) iodide is employed in the synthesis of chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 120°C (decomposition)
Physical Form Crystalline
Quantity 1 g
UN Number UN3260
Solubility Information Insoluble in water; Soluble in alkali iodide, cyanide solutions; Decomposes in warm acids
Formula Weight 323.87
Percent Purity 99%
Sensitivity Moisture sensitive
Density 8.25 g/mL
Chemical Name or Material Gold(I) iodide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'