missing translation for 'onlineSavingsMsg'
Learn More
Gold(I) sodium thiosulfate hydrate, 99.9% (metals basis), Thermo Scientific Chemicals Product Code.: 11374707

Gold(I) sodium thiosulfate hydrate, 99.9% (metals basis), Thermo Scientific Chemicals

Product Code. 11374707
25 g
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy

Product Code. 11374707

Brand: Thermo Scientific Alfa Aesar 039741.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Gold(I) sodium thiosulfate hydrate is useful in organogold compounds. Further, it serves as an antirheumatic agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15283-45-1
Molecular Formula AuNa3O6S4
Molecular Weight (g/mol) 490.17
MDL Number MFCD00046176
InChI Key FJCMCHNDCIYVST-UHFFFAOYSA-J
Synonym sodium aurothiosulfate, sodium gold thiosulfate, aurothiosulfate, sanochrysin, sanocrysine, crition, crytion, sanocrysin, aurate 3-, bis monothiosulfato-, trisodium, bis monothiosulfato 2--o,s aurate 3-trisodium t-4Show MoreShow Less
PubChem CID 27176
IUPAC Name gold(1+) trisodium disulfanidesulfonate
SMILES [Na+].[Na+].[Na+].[Au+].[O-]S([S-])(=O)=O.[O-]S([S-])(=O)=O

Specifications

Melting Point (decomposition)
Color White
Physical Form Powder
Quantity 25 g
Assay Percent Range 99.9% (Metal basis)
Solubility Information Soluble in water. Insoluble in ethanol and most other organic solvents.
Formula Weight 490.19 (anhydrous)
Appearance White powder
Density 3.09
Chemical Name or Material Gold(I) sodium thiosulfate hydrate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberW07K019 Certificate TypeCertificate of Analysis Date23/09/2024 Catalog Number039741.03,039741.14,039741.06
Lot NumberU02K025 Certificate TypeCertificate of Analysis Date16/08/2024 Catalog Number039741.03,039741.14,039741.06
Lot NumberR10K032 Certificate TypeCertificate of Analysis Date11/06/2024 Catalog Number039741.03,039741.14,039741.06
Lot NumberP02K011 Certificate TypeCertificate of Analysis Date18/04/2024 Catalog Number039741.03,039741.14,039741.06
Lot NumberQ09K064 Certificate TypeCertificate of Analysis Date15/04/2024 Catalog Number039741.03,039741.14,039741.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Gold(I) sodium thiosulfate hydrate, 99.9% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.