missing translation for 'onlineSavingsMsg'
Learn More

Gold(III) acetate, 99.9% (metals basis)

Product Code. 11304717
Click to view available options
Quantity:
0.5 g
1 g
5 g
Unit Size:
0.5g
1g
5g
This item is not returnable. View return policy

Product Code. 11304717

Brand: Thermo Scientific Alfa Aesar 039742.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 15804-32-7 | C6H9AuO6 | 374.099 g/mol

Used in solar energy technologies. Used in petrochemical cracking and automotive catalysts, water treatment, plating, textiles, research and in optic, laser, crystal and glass applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15804-32-7
Molecular Formula C6H9AuO6
Molecular Weight (g/mol) 374.099
MDL Number MFCD00210614
InChI Key OTCKNHQTLOBDDD-UHFFFAOYSA-K
Synonym gold 3+ triacetate, gold iii acetate, gold acetate, acmc-1c3i2, triacetic acid gold iii salt, acetic acid, gold 3+ salt 3:1
PubChem CID 15952561
IUPAC Name gold(3+);triacetate
SMILES CC(=O)[O-].CC(=O)[O-].CC(=O)[O-].[Au+3]

Specifications

Color Brown
Physical Form Powder
Quantity 5 g
Assay Percent Range 99.9% (Metal basis)
Solubility Information Slightly soluble in water.
Formula Weight 374.1
Appearance Brown powder
Chemical Name or Material Gold(III) acetate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.