missing translation for 'onlineSavingsMsg'
Learn More

Gold(III) chloride, 99.99+%, (trace metal basis), Thermo Scientific Chemicals

£206.03 - £834.15

Chemical Identifiers

CAS 13453-07-1
Molecular Formula AuCl3
Molecular Weight (g/mol) 303.32
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10047649
View Documents
Thermo Scientific Acros
437080010
1 g Glass bottle
£206.03
1g
Please to purchase this item. Need a web account? Register with us today!
15159293
View Documents
Thermo Scientific Acros
437080050
5 g Glass bottle
£834.15
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ultra trace analysis
Specifications

Chemical Identifiers

13453-07-1
AuCl3
303.32

Specifications

254.0°C
Brown to Red
63.0 to 66.9% (Au)
AuCl3
Solubility in water: soluble
303.32
Trace Metal Basis
13453-07-1
Crystalline Powder and/or Crystals
AuCl3
15,4556
303.32
≥99.99%
Gold(III) chloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Warning

missing translation for 'einecsNumber' : 236-623-1

Product Identifier
  • Gold(III) chloride

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contains : Gold chloride

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only