missing translation for 'onlineSavingsMsg'
Learn More

Gold(III) chloride, 99%, Thermo Scientific Chemicals

£105.53 - £834.15

Chemical Identifiers

CAS 13453-07-1
Molecular Formula AuCl3
Molecular Weight (g/mol) 303.32
MDL Number MFCD00014172
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10368980
View Documents
Thermo Scientific Acros
318645000
500 mg Glass bottle
£105.53
500mg
Please to purchase this item. Need a web account? Register with us today!
10135263
View Documents
Thermo Scientific Acros
318640050
5 g Glass bottle
£834.15
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

13453-07-1
AuCl3
303.32
MFCD00014172

Specifications

254.0°C
Brown to Yellow
98.5% min. (ex Au)
AuCl3
15, 4556
303.32
99%
13453-07-1
Crystalline Powder, Crystals and/or Chunks
AuCl3
MFCD00014172
Solubility in water: soluble
303.32
Gold(III) chloride, 99%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Warning

missing translation for 'einecsNumber' : 236-623-1

missing translation for 'rtecsNumber' : MD5420000

missing translation for 'tsca' : TSCA

Product Identifier
  • Gold(III) chloride

Signal Word
  • Warning

Hazard Category
  • Specific target organ toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2

Hazard Statement
  • H335-May cause respiratory irritation.
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only