missing translation for 'onlineSavingsMsg'
Learn More

Gold(III) chloride, 99%

Product Code. 10368980 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
500 mg
5 g
Packaging:
Glass bottle
Unit Size:
500mg
5g
This item is not returnable. View return policy

Product Code. 10368980

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13453-07-1 | AuCl3 | 303.32 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13453-07-1
Molecular Formula AuCl3
Molecular Weight (g/mol) 303.32
MDL Number MFCD00014172

Specifications

Melting Point 254.0°C
Color Brown to Yellow
Physical Form Crystalline Powder, Crystals and/or Chunks
Quantity 500 mg
Assay Percent Range 98.5% min. (ex Au)
Linear Formula AuCl3
Merck Index 15, 4556
Solubility Information Solubility in water: soluble
Formula Weight 303.32
Percent Purity 99%
Packaging Glass bottle
Chemical Name or Material Gold(III) chloride, 99%
Show More Show Less
compliance-icons
Product Identifier
  • Gold(III) chloride
Signal Word
  • Warning
Hazard Category
  • Specific target organ toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H335-May cause respiratory irritation.
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.