missing translation for 'onlineSavingsMsg'
Learn More

Gold(III) chloride trihydrate, Honeywell Fluka™

Product Code. 15667880
Click to view available options
Quantity:
1 g
5 g
10 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

ACS Reagent,≥49.0% Au basis

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16961-25-4
Molecular Formula AuCl4
Molecular Weight (g/mol) 338.77
MDL Number MFCD00149904
InChI Key CQVDKGFMVXRRAI-UHFFFAOYSA-J
IUPAC Name tetrachlorogold
SMILES Cl[Au](Cl)(Cl)Cl

Specifications

Quantity 25 g
Linear Formula HAuCl4 · 3H2O
UN Number UN3260
Formula Weight 393.83g/mol
Percent Purity ≥49% (Au basis)
Chemical Name or Material Gold(III) chloride trihydrate
compliance-icons
Product Identifier
  • Gold(III) chloride trihydrate
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'