missing translation for 'onlineSavingsMsg'
Learn More

Gold(III) hydroxide, Au 79% min

Product Code. 11366528
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
This item is not returnable. View return policy

Product Code. 11366528

Brand: Thermo Scientific Alfa Aesar 012159.09

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Gold plating solutions, porcelain decoration | CAS: 1303-52-2 | AuH6O3 | 251.01 g/mol

Gold(III) hydroxide is used in medicine, porcelain making, gold plating, and daguerrotypes. Used for preparation of gold catalysts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1303-52-2
Molecular Formula AuH6O3
Molecular Weight (g/mol) 251.01
MDL Number MFCD00046173
InChI Key PPOYUERUQZXZBE-UHFFFAOYSA-N
Synonym gold iii hydroxide, gold trihydroxide, gold hydroxide au oh 3, auh3o3, gold hydroxide, au oh 3, gold iii hydroxide, au
PubChem CID 102103
SMILES O.O.O.[Au]

Specifications

Physical Form Powder
Quantity 10 g
Merck Index 14,4523
Solubility Information Insoluble in water.
IUPAC Name gold;trihydrate
Formula Weight 247.99
Concentration or Composition (by Analyte or Components) Au 79% min.
Odor Odorless
Sensitivity Light sensitive
Chemical Name or Material Gold(III) hydroxide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.