Learn More
Gold nanoparticles, 10nm, supplied in 0.1mM PBS, reactant-free, 99%, OD1, 520nm absorption, Thermo Scientific Chemicals
£90.95 - £269.34
Chemical Identifiers
CAS | 7440-57-5 |
---|---|
Molecular Formula | Au |
Molecular Weight (g/mol) | 196.97 |
MDL Number | MFCD00003436 |
InChI Key | PCHJSUWPFVWCPO-UHFFFAOYSA-N |
Synonym | colloidal, powder, flake, leaf, gold, colloidal, burnish, shell, ci pigment metal 3, magnesium purple, c.i. pigment metal 3 |
PubChem CID | 23985 |
ChEBI | CHEBI:30050 |
IUPAC Name | gold |
SMILES | [Au] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15454565
|
Thermo Scientific Alfa Aesar
J67238.AC |
25 mL |
£90.95
25mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15464565
|
Thermo Scientific Alfa Aesar
J67238.AE |
100 mL |
£269.34
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Ideal for sensitive applications requiring a minimum amount of contaminating reactants. Well suited for protein conjugate development by passive adsorption to the gold nanoparticle surface. Conjugates can be used in blotting, electron microscopy and immunohistochemistry applications
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7440-57-5 | |
196.97 | |
PCHJSUWPFVWCPO-UHFFFAOYSA-N | |
23985 | |
gold |
Au | |
MFCD00003436 | |
colloidal, powder, flake, leaf, gold, colloidal, burnish, shell, ci pigment metal 3, magnesium purple, c.i. pigment metal 3 | |
CHEBI:30050 | |
[Au] |
Specifications
Red | |
Au | |
colloidal, powder, flake, leaf, gold, colloidal, burnish, shell, ci pigment metal 3, magnesium purple, c.i. pigment metal 3 | |
[Au] | |
196.97 | |
CHEBI:30050 | |
10nm | |
Liquid |
7440-57-5 | |
MFCD00003436 | |
PCHJSUWPFVWCPO-UHFFFAOYSA-N | |
gold | |
23985 | |
99% | |
Light sensitive | |
Gold nanoparticles, supplied in 0.1mM PBS, reactant-free, 99%, OD1, 520nm absorption |
Safety and Handling
missing translation for 'tsca' : Yes
missing translation for 'storageNote' : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only